Learn More
Strem, An Ascensus Company CAS# 185913-97-7. 250mg. (R)-(+)-5,5'-Dichloro-6,6'-dimethoxy-2,2'-bis(diphenylphosphino)-1,1'-biphenyl, min. 95% (R)-Cl-MeO-BIPHEP. MFCD03426987
SDP
Supplier:  Strem, An Ascensus Company 151055250MG
Encompass
CAS# 185913-97-7. 250mg. (R)-(+)-5,5'-Dichloro-6,6'-dimethoxy-2,2'-bis(diphenylphosphino)-1,1'-biphenyl, min. 95% (R)-Cl-MeO-BIPHEP. MFCD03426987. Molecular Weight: 651.50. Molecular Formula: C38H30Cl2O2P2. Color/form: yellow-white pwdr. Strem# 15-1055. http://www.strem.com/catalog/v/15-1055/
Catalog No. 50-901-10056