Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-5,7-Dimethoxyflavanone | 1277188-85-8 | 5 MG
SDP

Catalog No. 5000420749
Encompass_Preferred

(R)-5,7-Dimethoxyflavanone exhibits potent antimutagenic activity against MeIQ mutagenesis in Ames test utilizing S. typhimurium TA100 and TA98 strains. Additionally, it significantly and dose-dependently inhibits inflammatory mediators.

  • Potent antimutagenic activity
  • Inhibits inflammatory mediators
  • Molecular weight of 284.31
  • Chemical formula C17H16O4
  • Appears as a white to off-white solid
  • Classified as a flavonoid and flavanone
  • Initially sourced from plants

Catalog No. 50-004-20749 Supplier Medchemexpress LLC Supplier No. HYN50545MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R)-5,7-Dimethoxyflavanone exhibits potent antimutagenic activity against MeIQ mutagenesis in Ames test utilizing S. typhimurium TA100 and TA98 strains. Additionally, it significantly and dose-dependently inhibits inflammatory mediators.

  • Potent antimutagenic activity
  • Inhibits inflammatory mediators
  • Molecular weight of 284.31
  • Chemical formula C17H16O4
  • Appears as a white to off-white solid
  • Classified as a flavonoid and flavanone
  • Initially sourced from plants

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.