Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-5,7-Dimethoxyflavanone | 1277188-85-8 | MFCD31746978 | 1 MG
SDP

Catalog No. 5000419455
Encompass_Preferred

(R)-5,7-Dimethoxyflavanone is a compound that demonstrates potent antimutagenic activity against MeIQ mutagenesis in Ames tests using S. typhimurium TA100 and TA98 strains. It also significantly and dose-dependently inhibits inflammatory mediators. This product is for research use only.

  • Potent antimutagenic activity
  • Inhibits inflammatory mediators
  • Molecular weight: 284.31
  • Formula: C17H16O4
  • Appearance: solid
  • Color: white to off-white
  • Classified under flavonoids and flavanones
  • Initial source from plants

Catalog No. 50-004-19455 Supplier Medchemexpress LLC Supplier No. HYN50541MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R)-5,7-Dimethoxyflavanone is a compound that demonstrates potent antimutagenic activity against MeIQ mutagenesis in Ames tests using S. typhimurium TA100 and TA98 strains. It also significantly and dose-dependently inhibits inflammatory mediators. This product is for research use only.

  • Potent antimutagenic activity
  • Inhibits inflammatory mediators
  • Molecular weight: 284.31
  • Formula: C17H16O4
  • Appearance: solid
  • Color: white to off-white
  • Classified under flavonoids and flavanones
  • Initial source from plants

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.