Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-5-O-Benzoyl-1,2-di-O-isopropylidene-alpha-D-xylofuranose | 6612-91-5 | 95.1% | 294.30 | 1 ML
SDP

Catalog No. 5000403603
Encompass_Preferred

(R)-5-O-Benzoyl-1,2-di-O-isopropylidene-alpha-D-xylofuranose is a purine nucleoside analog. Purine nucleoside analogs exhibit broad antitumor activity, specifically targeting indolent lymphoid malignancies. The anticancer mechanisms associated with this process include inhibition of DNA synthesis and induction of apoptosis.

  • Appearance: solid
  • Color: white to off-white
  • Shipping: room temperature in continental US; may vary elsewhere
  • Storage: powder: -20°C for 3 years, 4°C for 2 years; in solvent: -80°C for 6 months, -20°C for 1 month
  • Purine nucleoside analog
  • Broad antitumor activity
  • Targets indolent lymphoid malignancies
  • Inhibits DNA synthesis
  • Induces apoptosis

Catalog No. 50-004-03603 Supplier Medchemexpress LLC Supplier No. HY4342610MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R)-5-O-Benzoyl-1,2-di-O-isopropylidene-alpha-D-xylofuranose is a purine nucleoside analog. Purine nucleoside analogs exhibit broad antitumor activity, specifically targeting indolent lymphoid malignancies. The anticancer mechanisms associated with this process include inhibition of DNA synthesis and induction of apoptosis.

  • Appearance: solid
  • Color: white to off-white
  • Shipping: room temperature in continental US; may vary elsewhere
  • Storage: powder: -20°C for 3 years, 4°C for 2 years; in solvent: -80°C for 6 months, -20°C for 1 month
  • Purine nucleoside analog
  • Broad antitumor activity
  • Targets indolent lymphoid malignancies
  • Inhibits DNA synthesis
  • Induces apoptosis

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.