Learn More
Medchemexpress LLC (R)-5-Oxopyrrolidine-2-carboxylic acid | 4042-36-8 | 129.11 | 5 G
SDP
Catalog No. 5000445621
Encompass_Preferred
(R)-5-Oxopyrrolidine-2-carboxylic acid (D-Pyroglutamic acid) is a cyclic derivative of D-glutamate. This compound is suitable for research pertaining to metabolic syndrome and end-stage renal disease.
  • Cyclic derivative of D-glutamate
  • Utilized in research pertaining to metabolic syndrome and end-stage renal disease
  • Appears as a white to off-white solid
  • Classified as a ketone, aldehyde, and acid
  • Functions as an endogenous metabolite
Catalog No. 50-004-45621 Supplier Medchemexpress LLC Supplier No. HYW0023045G
(R)-5-Oxopyrrolidine-2-carboxylic acid (D-Pyroglutamic acid) is a cyclic derivative of D-glutamate. This compound is suitable for research pertaining to metabolic syndrome and end-stage renal disease.
  • Cyclic derivative of D-glutamate
  • Utilized in research pertaining to metabolic syndrome and end-stage renal disease
  • Appears as a white to off-white solid
  • Classified as a ketone, aldehyde, and acid
  • Functions as an endogenous metabolite