Learn More
Medchemexpress LLC HY-107613 5mg Medchemexpress, R 59-022 CAS:93076-89-2 Purity:>98%
SDP
Catalog No. 501968439
Encompass_Preferred
Medchemexpress, HY-107613 5mg R 59-022 CAS:93076-89-2 R 59-022 (DKGI-I) is a diacylglycerol kinase inhibitor (IC50=2.8 μM). R 59-022 is a 5-HTR antagonist, and activates protein kinase C (PKC). R 59-022 potentiates thrombin-induced diacylglycerol production in platelets and inhibits phosphatidic acid production in neutrophils[3][4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8439 Supplier Medchemexpress LLC Supplier No. HY1076135MG
Medchemexpress, HY-107613 5mg R 59-022 CAS:93076-89-2 R 59-022 (DKGI-I) is a diacylglycerol kinase inhibitor (IC50=2.8 μM). R 59-022 is a 5-HTR antagonist, and activates protein kinase C (PKC). R 59-022 potentiates thrombin-induced diacylglycerol production in platelets and inhibits phosphatidic acid production in neutrophils[3][4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.