Learn More
Apexbio Technology LLC R 59-022 93076-89-2 10mg
SDP
Catalog No. 5000090254
Encompass_Preferred
R 59-022 (CAS 93076-89-2) is a small molecule inhibitor of diacylglycerol kinase (DGK) demonstrating an IC50 of 2 8 M for DGK activity By inhibiting the conversion of 1-oleoyl-2-acetylglycerol to phosphatidic acid R 59-022 modulates diacylglycerol signaling pathways The compound also exhibits antagonistic effects at serotonin (5-HT) receptors and can induce activation of protein kinase C (PKC) In cellular studies R 59-022 enhances thrombin-induced diacylglycerol accumulation in platelets and suppresses phosphatidic acid formation in neutrophils It serves as a valuable tool for investigating DGK-mediated processes and signal transduction mechanisms
Catalog No. 50-000-90254 Supplier Apexbio Technology LLC Supplier No. B696710
R 59-022 (CAS 93076-89-2) is a small molecule inhibitor of diacylglycerol kinase (DGK) demonstrating an IC50 of 2 8 M for DGK activity By inhibiting the conversion of 1-oleoyl-2-acetylglycerol to phosphatidic acid R 59-022 modulates diacylglycerol signaling pathways The compound also exhibits antagonistic effects at serotonin (5-HT) receptors and can induce activation of protein kinase C (PKC) In cellular studies R 59-022 enhances thrombin-induced diacylglycerol accumulation in platelets and suppresses phosphatidic acid formation in neutrophils It serves as a valuable tool for investigating DGK-mediated processes and signal transduction mechanisms