Learn More
Apexbio Technology LLC R 59-022 93076-89-2 5mg
SDP
Numéro de catalogue. 5000089769
Encompass_Preferred
R 59-022 (CAS 93076-89-2) is a small molecule inhibitor of diacylglycerol kinase (DGK) demonstrating an IC50 of 2 8 M for DGK activity By inhibiting the conversion of 1-oleoyl-2-acetylglycerol to phosphatidic acid R 59-022 modulates diacylglycerol signaling pathways The compound also exhibits antagonistic effects at serotonin (5-HT) receptors and can induce activation of protein kinase C (PKC) In cellular studies R 59-022 enhances thrombin-induced diacylglycerol accumulation in platelets and suppresses phosphatidic acid formation in neutrophils It serves as a valuable tool for investigating DGK-mediated processes and signal transduction mechanisms
Numéro de catalogue. 50-000-89769 Fournisseur Apexbio Technology LLC Code fournisseur B69675
Ajouter au panier
Ajouter au panier
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
R 59-022 (CAS 93076-89-2) is a small molecule inhibitor of diacylglycerol kinase (DGK) demonstrating an IC50 of 2 8 M for DGK activity By inhibiting the conversion of 1-oleoyl-2-acetylglycerol to phosphatidic acid R 59-022 modulates diacylglycerol signaling pathways The compound also exhibits antagonistic effects at serotonin (5-HT) receptors and can induce activation of protein kinase C (PKC) In cellular studies R 59-022 enhances thrombin-induced diacylglycerol accumulation in platelets and suppresses phosphatidic acid formation in neutrophils It serves as a valuable tool for investigating DGK-mediated processes and signal transduction mechanisms
missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.