Learn More
Medchemexpress LLC R 59-022 hydrochlori 10mg | 93076-98-3 | 10MG
SDP
Catalog No. 5000240848
Encompass_Preferred
R 59-022 (DKGI-I) hydrochloride is a DGK inhibitor (IC50 2 8 M) R 59-022 hydrochloride inhibits the phosphorylation of OAG to OAPA R 59-022 hydrochloride is a 5-HT Receptor antagonist and activates protein kinase C (PKC) R 59-022 hydrochloride potentiates thrombin-induced diacylglycerol production in platelets and inhibits phosphatidic acid production in neutrophils[1 [2 [3 [4
Catalog No. 50-002-40848 Supplier Medchemexpress LLC Supplier No. HY107613A10MG
R 59-022 (DKGI-I) hydrochloride is a DGK inhibitor (IC50 2 8 M) R 59-022 hydrochloride inhibits the phosphorylation of OAG to OAPA R 59-022 hydrochloride is a 5-HT Receptor antagonist and activates protein kinase C (PKC) R 59-022 hydrochloride potentiates thrombin-induced diacylglycerol production in platelets and inhibits phosphatidic acid production in neutrophils[1 [2 [3 [4