Learn More
TARGETMOL CHEMICALS INC R 59-022 HYDROCHLORIDE 5MG
SDP
Catalog No. 502579233
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. R 59-022 hydrochloride (DKGI-I hydrochloride) is a 5-HT Receptor antagonist that activates protein kinase C (PKC).R 59-022 hydrochloride is a DGK inhibitor (IC502.8 µM) that inhibits the phosphorylation of OAG to OAPA.R 59-022 enhances thrombin-induced triglyceride production in platelets and inhibits the production of phosphatidic acid in neutrophils. R 59-022 enhances thrombin-induced diglyceride production in platelets and inhibits phosphatidic acid production in neutrophils. purity: 97%
Catalog No. 50-257-9233 Supplier TARGETMOL CHEMICALS INC Supplier No. T723225MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. R 59-022 hydrochloride (DKGI-I hydrochloride) is a 5-HT Receptor antagonist that activates protein kinase C (PKC).R 59-022 hydrochloride is a DGK inhibitor (IC502.8 µM) that inhibits the phosphorylation of OAG to OAPA.R 59-022 enhances thrombin-induced triglyceride production in platelets and inhibits the production of phosphatidic acid in neutrophils. R 59-022 enhances thrombin-induced diglyceride production in platelets and inhibits phosphatidic acid production in neutrophils. purity: 97%