Learn More
Medchemexpress LLC R 59-022 (hydrochloride) | 93076-98-3 | MFCD34676749 | 98.9% | 496.04 | C27H27ClFN3OS | 5 MG
SDP
Catalog No. 5000240049
Encompass_Preferred
R 59-022 (hydrochloride) is a research-grade small-molecule inhibitor used to probe diacylglycerol kinase (DGK) activity and downstream lipid signaling pathways in biochemical and cellular studies. The compound is supplied as a hydrochloride salt with reported molecular weight 496.04 and high analytical purity appropriate for laboratory research applications.
  • Selective diacylglycerol kinase inhibitor activity.
  • Potentiates protein kinase C activation in cellular assays.
  • High purity suitable for biochemical and cell-based studies.
  • Supplied as a hydrochloride salt to aid solubility in polar solvents.
  • Provided in small research pack sizes for laboratory use.
Catalog No. 50-002-40049 Supplier Medchemexpress LLC Supplier No. HY107613A5MG
R 59-022 (hydrochloride) is a research-grade small-molecule inhibitor used to probe diacylglycerol kinase (DGK) activity and downstream lipid signaling pathways in biochemical and cellular studies. The compound is supplied as a hydrochloride salt with reported molecular weight 496.04 and high analytical purity appropriate for laboratory research applications.
  • Selective diacylglycerol kinase inhibitor activity.
  • Potentiates protein kinase C activation in cellular assays.
  • High purity suitable for biochemical and cell-based studies.
  • Supplied as a hydrochloride salt to aid solubility in polar solvents.
  • Provided in small research pack sizes for laboratory use.