Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC R 59-022 (hydrochloride) | 93076-98-3 | MFCD34676749 | 98.9% | 496.04 g/mol | C27H27ClFN3OS | 25 MG
SDP

Catalog No. 5000240133
Encompass_Preferred

R 59-022 (hydrochloride) is the hydrochloride salt of a small-molecule diacylglycerol kinase inhibitor used for laboratory research. It is provided as an off-white to light yellow solid with molecular formula C27H27ClFN3OS and molecular weight 496.04 g/mol. The compound is reported with high purity and is accompanied by an SDS for safe handling and storage.

  • Used as a diacylglycerol kinase inhibitor in biochemical assays.
  • High purity (98.9%).
  • Molecular weight 496.04 g/mol.
  • Formula C27H27ClFN3OS.
  • Off-white to light yellow solid appearance for easy visual inspection.
  • Supplied in 25 mg research quantities suitable for screening and assay development.

Catalog No. 50-002-40133 Supplier Medchemexpress LLC Supplier No. HY107613A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

R 59-022 (hydrochloride) is the hydrochloride salt of a small-molecule diacylglycerol kinase inhibitor used for laboratory research. It is provided as an off-white to light yellow solid with molecular formula C27H27ClFN3OS and molecular weight 496.04 g/mol. The compound is reported with high purity and is accompanied by an SDS for safe handling and storage.

  • Used as a diacylglycerol kinase inhibitor in biochemical assays.
  • High purity (98.9%).
  • Molecular weight 496.04 g/mol.
  • Formula C27H27ClFN3OS.
  • Off-white to light yellow solid appearance for easy visual inspection.
  • Supplied in 25 mg research quantities suitable for screening and assay development.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.