Learn More
Medchemexpress LLC R 59-022 (hydrochloride) | 93076-98-3 | MFCD34676749 | 98.9% | 496.04 g·mol⁻¹ | C27H27ClFN3OS | 50 MG
SDP
Catalog No. 5000241480
Encompass_Preferred
R 59-022 hydrochloride is the hydrochloride salt of R 59-022, a research compound that acts as a diacylglycerol kinase (DGK) inhibitor (reported IC50 ≈ 2.8 μM). It is used in cellular and biochemical assays to modulate diacylglycerol/protein kinase C signaling and to probe 5-HT receptor-related pathways. The material is supplied as an off-white solid (CAS 93076-98-3) with formula C27H27ClFN3OS and molecular weight 496.04 g·mol⁻¹.
  • DGK inhibitor with reported IC50 ≈ 2.8 μM.
  • Modulates protein kinase C and diacylglycerol signaling in cells.
  • Hydrochloride salt form for improved handling and solubility in polar solvents.
  • High purity suitable for research applications.
  • Soluble in DMSO for preparation of stock solutions.
Catalog No. 50-002-41480 Supplier Medchemexpress LLC Supplier No. HY107613A50MG
R 59-022 hydrochloride is the hydrochloride salt of R 59-022, a research compound that acts as a diacylglycerol kinase (DGK) inhibitor (reported IC50 ≈ 2.8 μM). It is used in cellular and biochemical assays to modulate diacylglycerol/protein kinase C signaling and to probe 5-HT receptor-related pathways. The material is supplied as an off-white solid (CAS 93076-98-3) with formula C27H27ClFN3OS and molecular weight 496.04 g·mol⁻¹.
  • DGK inhibitor with reported IC50 ≈ 2.8 μM.
  • Modulates protein kinase C and diacylglycerol signaling in cells.
  • Hydrochloride salt form for improved handling and solubility in polar solvents.
  • High purity suitable for research applications.
  • Soluble in DMSO for preparation of stock solutions.