Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(-)-6,6'-Dibromo-1,1'-bi-2-naphthol 99%, Thermo Scientific™

Catalog No. AC333965000
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC333965000 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC333965000 Supplier Thermo Scientific Chemicals Supplier No. 333965000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65283-60-5
Molecular Formula C20H12Br2O2
Molecular Weight (g/mol) 444.122
InChI Key OORIFUHRGQKYEV-UHFFFAOYSA-N
Synonym r---6,6'-dibromo-1,1'-bi-2-naphthol, s---6,6'-dibromo-1,1'-bi-2-naphthol, s-+-6,6'-dibromo-1,1'-bi-2-naphthol, 6,6'-dibromo-1,1'-bi-2-naphthol, s-+-6,6'-dibromo-1,1'-binaphthalene-2,2'-diol, r---6,6'-dibromo-1,1'-binaphthalene-2,2'-diol, 6,6'-dibromo 1,1'-binaphthalene-2,2'-diol, +/--6,6'-dibromo-1,1'-bi-2-naphthol, s-6,6'-dibromo-1,1'-bi-2-naphthol, racemic-6,6'-dibromo-1,1'-bi-2-naphthol
PubChem CID 222842
IUPAC Name 6-bromo-1-(6-bromo-2-hydroxynaphthalen-1-yl)naphthalen-2-ol
SMILES C1=CC2=C(C=CC(=C2C3=C(C=CC4=C3C=CC(=C4)Br)O)O)C=C1Br

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Green to White
Infrared Spectrum Authentic
Assay Percent Range 99%
Quantity 500 mg
Specific Rotation Condition − 48.00 (20.00°C c=1.8,THF)
Specific Rotation -46° to -52° (20°C, 589nm) (c=1.8, THF)
Formula Weight 444.12
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material (R)-(-)-6, 6'-Dibromo-1, 1'-bi-2-naphthol, 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.