Learn More
Medchemexpress LLC (R)-6-chloro-N-(4-fluoro-2-(methylsulfonyl)phenyl)-7-hydroxy-4-oxo-4H-chromene-2-carboxamide | 2438637-65-9 | 99.8% | 435.81 g/mol | C19H11ClFNO6S | 5 MG
SDP
Catalog No. 5000262391
Encompass_Preferred
PGAM1-IN-1 is a small-molecule inhibitor of phosphoglycerate mutase 1 (PGAM1) with an enzymatic IC50 of 6.4 μM and demonstrated cellular activity (H1299 proliferation IC50 14.1 ± 1.9 μM). Supplied as a high-purity research reagent, the compound has molecular formula C19H11ClFNO6S and molecular weight 435.81 g/mol. For research use only.
  • Inhibits phosphoglycerate mutase 1 with an IC50 of 6.4 μM.
  • Shows H1299 cell proliferation inhibition (IC50 14.1 ± 1.9 μM).
  • High purity (>99.8%).
  • Available as solid and as a 10 mM solution in DMSO.
  • Molecular formula C19H11ClFNO6S; molecular weight 435.81 g/mol.
Catalog No. 50-002-62391 Supplier Medchemexpress LLC Supplier No. HY1286815MG
PGAM1-IN-1 is a small-molecule inhibitor of phosphoglycerate mutase 1 (PGAM1) with an enzymatic IC50 of 6.4 μM and demonstrated cellular activity (H1299 proliferation IC50 14.1 ± 1.9 μM). Supplied as a high-purity research reagent, the compound has molecular formula C19H11ClFNO6S and molecular weight 435.81 g/mol. For research use only.
  • Inhibits phosphoglycerate mutase 1 with an IC50 of 6.4 μM.
  • Shows H1299 cell proliferation inhibition (IC50 14.1 ± 1.9 μM).
  • High purity (>99.8%).
  • Available as solid and as a 10 mM solution in DMSO.
  • Molecular formula C19H11ClFNO6S; molecular weight 435.81 g/mol.