Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-7-cyano-2,3,4,5-tetrahydro-1-(1H-imidazol-4-ylmethyl)-3-(phenylmethyl)-4-(2-thienylsulfonyl) | 195987-41-8 | 99.9% | 489.61 g/mol | C25H23N5O2S2 | 10 MG
SDP

Catalog No. 5000288530
Encompass_Preferred

A small-molecule farnesyl transferase inhibitor supplied as a high-purity solid for research use. It inhibits prenylation of Ras family proteins and is intended for laboratory studies requiring a selective FTase inhibitor.

  • High purity (99.9%)
  • Solid form, suitable for formulation or dissolution
  • Molecular weight 489.61 g/mol
  • Chemical formula C25H23N5O2S2
  • Recommended storage: powder -20°C for long term; solvent storage guidelines available
  • Available as a 10 mg research pack

Catalog No. 50-002-88530 Supplier Medchemexpress LLC Supplier No. HY1611110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A small-molecule farnesyl transferase inhibitor supplied as a high-purity solid for research use. It inhibits prenylation of Ras family proteins and is intended for laboratory studies requiring a selective FTase inhibitor.

  • High purity (99.9%)
  • Solid form, suitable for formulation or dissolution
  • Molecular weight 489.61 g/mol
  • Chemical formula C25H23N5O2S2
  • Recommended storage: powder -20°C for long term; solvent storage guidelines available
  • Available as a 10 mg research pack

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.