Learn More
TARGETMOL CHEMICALS INC R-7050 25MG
SDP
Catalog No. 502579858
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. R-7050 (TNF-(alpha) Antagonist III) is a tumor necrosis factor receptor (TNFR) antagonist with greater selectivity toward TNF(alpha). purity: 99%
Catalog No. 50-257-9858 Supplier TARGETMOL CHEMICALS INC Supplier No. T463725MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. R-7050 (TNF-(alpha) Antagonist III) is a tumor necrosis factor receptor (TNFR) antagonist with greater selectivity toward TNF(alpha). purity: 99%