Learn More
Cayman Chemical R 715trIfluoroacetAT salt 10mg
SDP
Catalog No. 502915011
Encompass_Preferred
A bradykinin B1 receptor antagonist; inhibits bradykinin-induced contractions in isolated human umbilical cords, which express bradykinin B1 receptors (pA2 = 8.49); decreases the latency to tail withdrawal in the tail-flick test in an STZ-induced mouse model of diabetic neuropathy at 200, 400, and 600 UG/kg; reduces the incidence of hind limb weakness and paralysis, improves symmetrical gait, as well as decreases spinal inflammatory foci numbers, neuron demyelination, and lesion monocyte invasion, in a MOG35-55 peptide-induced mouse model of EAE at 1 mg/kg per day; reduces mean arterial blood pressure and increases heart rate in spontaneously hypertensive rats at 0.01 nmol/animal, i.c.v
Catalog No. 50-291-5011 Supplier Cayman Chemical Supplier No. 3689010 MG
A bradykinin B1 receptor antagonist; inhibits bradykinin-induced contractions in isolated human umbilical cords, which express bradykinin B1 receptors (pA2 = 8.49); decreases the latency to tail withdrawal in the tail-flick test in an STZ-induced mouse model of diabetic neuropathy at 200, 400, and 600 UG/kg; reduces the incidence of hind limb weakness and paralysis, improves symmetrical gait, as well as decreases spinal inflammatory foci numbers, neuron demyelination, and lesion monocyte invasion, in a MOG35-55 peptide-induced mouse model of EAE at 1 mg/kg per day; reduces mean arterial blood pressure and increases heart rate in spontaneously hypertensive rats at 0.01 nmol/animal, i.c.v