Learn More
Medchemexpress LLC (R)-8-(4-(trifluoromethyl)phenyl)-N-((S)-1,1,1-trifluoropropan-2-yl)-5,6-dihydro-1,7-naphthyridine | 1470018-52-0 | 99.9% | 417.35 g·mol⁻¹ | C19H17F6N3O | 25 MG
SDP
Catalog No. 5000244061
Encompass_Preferred
AMG2850 is a potent, orally bioavailable, and selective antagonist of the transient receptor potential melastatin 8 (TRPM8) ion channel, provided as a research reagent for preclinical in vitro and in vivo studies (CAS 1470018-52-0).
  • Potent, selective TRPM8 antagonist for target pharmacology.
  • Demonstrated oral bioavailability in preclinical models.
  • High reported purity (99.9%).
  • Available as solid powder or as 10 mM solution in DMSO.
  • Molecular weight 417.35 g·mol⁻¹ and formula C19H17F6N3O.
  • For research use only; not for human or veterinary applications.
Catalog No. 50-002-44061 Supplier Medchemexpress LLC Supplier No. HY10405925MG
AMG2850 is a potent, orally bioavailable, and selective antagonist of the transient receptor potential melastatin 8 (TRPM8) ion channel, provided as a research reagent for preclinical in vitro and in vivo studies (CAS 1470018-52-0).
  • Potent, selective TRPM8 antagonist for target pharmacology.
  • Demonstrated oral bioavailability in preclinical models.
  • High reported purity (99.9%).
  • Available as solid powder or as 10 mM solution in DMSO.
  • Molecular weight 417.35 g·mol⁻¹ and formula C19H17F6N3O.
  • For research use only; not for human or veterinary applications.