Learn More
Medchemexpress LLC R-8-(4-trifluoromethylphenyl)-N-(S-1,1,1-trifluoropropan-2-yl)-1,7-naphthyridine-7-carboxamide | 1470018-52-0 | 99.9% | 417.4 g·mol⁻1 | C19H17F6N3O | 50 MG
SDP
Catalog No. 5000241854
Encompass_Preferred
AMG2850 is a small-molecule, selective antagonist of the transient receptor potential melastatin 8 (TRPM8) ion channel. It is described in the literature as orally bioavailable and is used in preclinical research to probe TRPM8-mediated pharmacology. Identifiers include CAS 1470018-52-0, chemical formula C19H17F6N3O, and molecular weight ≈417.4 g·mol⁻1.
  • Selective TRPM8 antagonist for target validation in preclinical studies.
  • Orally bioavailable with reported in vivo exposure in rodent models.
  • Well characterized in peer-reviewed studies for pharmacology and selectivity.
  • High chemical purity suitable for research applications.
  • Provided with standard analytical identifiers for compound verification.
Catalog No. 50-002-41854 Supplier Medchemexpress LLC Supplier No. HY10405950MG
AMG2850 is a small-molecule, selective antagonist of the transient receptor potential melastatin 8 (TRPM8) ion channel. It is described in the literature as orally bioavailable and is used in preclinical research to probe TRPM8-mediated pharmacology. Identifiers include CAS 1470018-52-0, chemical formula C19H17F6N3O, and molecular weight ≈417.4 g·mol⁻1.
  • Selective TRPM8 antagonist for target validation in preclinical studies.
  • Orally bioavailable with reported in vivo exposure in rodent models.
  • Well characterized in peer-reviewed studies for pharmacology and selectivity.
  • High chemical purity suitable for research applications.
  • Provided with standard analytical identifiers for compound verification.