Learn More
Medchemexpress LLC (R)-8-hydroxy-2-dipropylaminotetralin hydrobromide | 78095-19-9 | MFCD00055158 | 99.3% | 328.29 g·mol⁻¹ | C16H26BrNO | 5 MG
SDP
Catalog No. 5000264266
Encompass_Preferred
R(+)-8-OH-DPAT hydrobromide is the R-enantiomer of 8-OH-DPAT, a potent and selective 5-HT1A receptor agonist widely used in neuroscience research to probe serotonergic function. Supplied as the hydrobromide salt, this compound is provided as a small-quantity, high-purity material for in vitro and in vivo studies.
  • Potent, selective 5-HT1A receptor agonist.
  • R-enantiomer for stereospecific pharmacology.
  • Hydrobromide salt form for improved solubility.
  • High purity (99.3%) suitable for research applications.
  • Molecular formula C16H26BrNO and molecular weight 328.29 g·mol⁻¹.
  • Available in small research quantities (5 MG).
Catalog No. 50-002-64266 Supplier Medchemexpress LLC Supplier No. HY101377A5MG
R(+)-8-OH-DPAT hydrobromide is the R-enantiomer of 8-OH-DPAT, a potent and selective 5-HT1A receptor agonist widely used in neuroscience research to probe serotonergic function. Supplied as the hydrobromide salt, this compound is provided as a small-quantity, high-purity material for in vitro and in vivo studies.
  • Potent, selective 5-HT1A receptor agonist.
  • R-enantiomer for stereospecific pharmacology.
  • Hydrobromide salt form for improved solubility.
  • High purity (99.3%) suitable for research applications.
  • Molecular formula C16H26BrNO and molecular weight 328.29 g·mol⁻¹.
  • Available in small research quantities (5 MG).