Learn More
Bellco Glass, Inc. Roller Apparatus Base & 2 Decks, 115V Modular - 15 Position
SDP
Catalog No. 501945916
Encompass_Preferred
The Bellco Cell Production Roller Apparatus has been designed for production of animal cells, viruses and cell products. Modular units are available from 5 to 45 bottle positions and allow easy expansion with modular add-on decks. Non-modular production units come in 45 and 50 bottle positions with over 80,000 cm2 of surface area. Rollers have a key design to eliminate slipping and are easily removable for servicing of bearings. Rollers are formulated from a chemical-resistant non-slip EDPM. All units feature a solid state control with heavy-duty DC motor and analog tachometer. Non-modular production models feature rubber bumpers, handles and a slotted rear guard. MADE IN THE USA
Catalog No. 50-194-5916 Supplier Bellco Glass, Inc. Supplier No. 763075012
The Bellco Cell Production Roller Apparatus has been designed for production of animal cells, viruses and cell products. Modular units are available from 5 to 45 bottle positions and allow easy expansion with modular add-on decks. Non-modular production units come in 45 and 50 bottle positions with over 80,000 cm2 of surface area. Rollers have a key design to eliminate slipping and are easily removable for servicing of bearings. Rollers are formulated from a chemical-resistant non-slip EDPM. All units feature a solid state control with heavy-duty DC motor and analog tachometer. Non-modular production models feature rubber bumpers, handles and a slotted rear guard. MADE IN THE USA