Learn More
Bellco Glass, Inc. Roller Apparatus Base & 6 Decks, 115V Modular - 35 Position
SDP
Catalog No. 501945920
Encompass_Preferred
The Bellco Cell Production Roller Apparatus has been designed for production of animal cells, viruses and cell products. Modular units are available from 5 to 45 bottle positions and allow easy expansion with modular add-on decks. Non-modular production units come in 45 and 50 bottle positions with over 80,000 cm2 of surface area. Rollers have a key design to eliminate slipping and are easily removable for servicing of bearings. Rollers are formulated from a chemical-resistant non-slip EDPM. All units feature a solid state control with heavy-duty DC motor and analog tachometer. Non-modular production models feature rubber bumpers, handles and a slotted rear guard. MADE IN THE USA
Catalog No. 50-194-5920 Supplier Bellco Glass, Inc. Supplier No. 763075016
Add to Cart
Add to Cart
Request a Quote
The Bellco Cell Production Roller Apparatus has been designed for production of animal cells, viruses and cell products. Modular units are available from 5 to 45 bottle positions and allow easy expansion with modular add-on decks. Non-modular production units come in 45 and 50 bottle positions with over 80,000 cm2 of surface area. Rollers have a key design to eliminate slipping and are easily removable for servicing of bearings. Rollers are formulated from a chemical-resistant non-slip EDPM. All units feature a solid state control with heavy-duty DC motor and analog tachometer. Non-modular production models feature rubber bumpers, handles and a slotted rear guard. MADE IN THE USA