Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical (R)-alpha-(Fmoc-amino)-3-hydroxy-benzeneacetic acid | 25 g | CAS 615553-60-1 | MDL MFCD07371748

Catalog No. 502345460
Encompass

(R)-alpha-(Fmoc-amino)-3-hydroxy-benzeneacetic acid is a Amino Acid reagent (Subcategory: Aryl AA) sold by WuXi TIDES. Offered in 25 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 615553-60-1
- MDL: MFCD07371748
- InChIKey: JAVDQXYAZCMANV-OAQYLSRUSA-N
- Molecular Weight: 389.407
- Molecular Formula: C23H19NO5
- Purity: ≥95%
- Container Type: 125 mL HDPE
- Pack Size: 25 g
- Net Weight: 25 g
- Gross Weight: 54.3 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: (R)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-2-(3-hydroxyphenyl)acetic acid
- SMILES: O=C(OCC1C2=CC=CC=C2C3=C1C=CC=C3)N[C@H](C4=CC(O)=CC=C4)C(O)=O

Catalog No. 50-234-5460 Supplier STA Pharmaceutical Supplier No. STAAA1163725GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R)-alpha-(Fmoc-amino)-3-hydroxy-benzeneacetic acid is a Amino Acid reagent (Subcategory: Aryl AA) sold by WuXi TIDES. Offered in 25 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 615553-60-1
- MDL: MFCD07371748
- InChIKey: JAVDQXYAZCMANV-OAQYLSRUSA-N
- Molecular Weight: 389.407
- Molecular Formula: C23H19NO5
- Purity: ≥95%
- Container Type: 125 mL HDPE
- Pack Size: 25 g
- Net Weight: 25 g
- Gross Weight: 54.3 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: (R)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-2-(3-hydroxyphenyl)acetic acid
- SMILES: O=C(OCC1C2=CC=CC=C2C3=C1C=CC=C3)N[C@H](C4=CC(O)=CC=C4)C(O)=O

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.