Learn More
Cayman Chemical R-a-MethylhIstamInhydr 10mg
SDP
Catalog No. 502936699
Encompass_Preferred
A histamine H3 receptor agonist; inhibits the release of histamine induced by antidromic electrical stimulation of the sciatic nerve in rats from 0.25-2 mg/kg; inhibits gastric acid secretion in rats when administered intracerebroventricularly from 0.5-50 nmol; reduces isolation-induced vocalizations and aggressive behavior in a resident-intruder test in guinea pig pups and mice, respectively; decreases freezing time in a conditioned fear stress test in rats; acts synergistically with fentanyl to reduce nociception and plasma extravasation in a mouse model of chronic inflammation induced by complete Freund's adjuvant
Catalog No. 50-293-6699 Supplier Cayman Chemical Supplier No. 2560110 MG
A histamine H3 receptor agonist; inhibits the release of histamine induced by antidromic electrical stimulation of the sciatic nerve in rats from 0.25-2 mg/kg; inhibits gastric acid secretion in rats when administered intracerebroventricularly from 0.5-50 nmol; reduces isolation-induced vocalizations and aggressive behavior in a resident-intruder test in guinea pig pups and mice, respectively; decreases freezing time in a conditioned fear stress test in rats; acts synergistically with fentanyl to reduce nociception and plasma extravasation in a mouse model of chronic inflammation induced by complete Freund's adjuvant