Learn More
Medchemexpress LLC HY-13058B 5mg Medchemexpress, (R)-ADX-47273 CAS:851881-59-9 Purity:>98%
SDP
Catalog No. 501932649
Encompass_Preferred
Medchemexpress, HY-13058B 5mg (R)-ADX-47273 CAS:851881-59-9 (R)-ADX-47273 is a potent mGluR5 positive allosteric modulator, with an EC 50 of 168 nM for potentiation . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2649 Supplier Medchemexpress LLC Supplier No. HY13058B5MG
Medchemexpress, HY-13058B 5mg (R)-ADX-47273 CAS:851881-59-9 (R)-ADX-47273 is a potent mGluR5 positive allosteric modulator, with an EC 50 of 168 nM for potentiation . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.