Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-alpha,alpha-Diphenyl-2-pyrrolidinemethanol Trimethylsilyl Ether 98.0+%, TCI America™
SDP

Catalog No. D38675G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38675G 5 g
D38671G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D38675G Supplier TCI America Supplier No. D38675G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 943757-71-9
Molecular Formula C20H27NOSi
Molecular Weight (g/mol) 325.53
MDL Number MFCD09265100
InChI Key RSUHWMSTWSSNOW-UHFFFAOYNA-N
Synonym (R)-(+)-2-[Diphenyl(trimethylsilyloxy)methyl]pyrrolidine, alpha,alpha-Diphenyl-D-prolinol Trimethylsilyl Ether, (R)-Hayashi-Jorgensen Catalyst
PubChem CID 16218310
IUPAC Name 2-{diphenyl[(trimethylsilyl)oxy]methyl}pyrrolidine
SMILES C[Si](C)(C)OC(C1CCCN1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 325.53
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material (R)-(+)-alpha,alpha-Diphenyl-2-pyrrolidinemethanol Trimethylsilyl Ether
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.