Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-alpha-Methyltryptophan hemihydrate, 98%, 98% ee

Catalog No. AC441191000
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC441191000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC441191000 Supplier Thermo Scientific Chemicals Supplier No. 441191000
Add to Cart

CAS: 56452-52-9 | C24H30N4O5 | 454.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56452-52-9
Molecular Formula C24H30N4O5
Molecular Weight (g/mol) 454.53
MDL Number MFCD19687046
InChI Key BZCFHAYHUKOGKE-IUWNWRFPSA-N
Synonym r-a-methyltryptophan, alpha-methyl-d-tryptophan, 2r-2-amino-3-1h-indol-3-yl-2-methylpropanoic acid, d-a-methyltryptophan, +-a-methyltryptophan, a-methyl-r-tryptophan, h-a-me-d-trp-oh, ncistruc1_000316, ncistruc2_000076, a-methyl-d-tryptophan
PubChem CID 676154
IUPAC Name (2R)-2-amino-3-(1H-indol-3-yl)-2-methylpropanoic acid
SMILES O.CC(N)(CC1=CNC2=CC=CC=C12)C(O)=O.CC(N)(CC1=CNC2=CC=CC=C12)C(O)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point >231°C
Color White to Yellow
Enantiomeric Excess 98.0% min.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 100 mg
Specific Rotation 16
Formula Weight 227.28
Percent Purity 98%,98% ee
Physical Form Powder
Chemical Name or Material (R)-α-Methyltryptophan hemihydrate, 98% ee
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.