Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich R-Alpine-Borane™

Catalog No. 41720325ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-720-325ML 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 41-720-325ML Supplier Sigma Aldrich Supplier No. 41720325ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

R-Alpine-Borane™ is a chiral reducing agent, synthesized from (+)-+a-pinene via hydroboration.Reagent for the asymmetric reduction of a variety of prochiral ketones.Alpine-Borane is a registered trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 1530-45-6
Melting Point 145°C to 150°C
Molecular Formula C22H22BrO2P
Linear Formula CH3CH2OCOCH2P(C6H5)3Br
MDL Number MFCD00011835
Quantity 100 g
Synonym B-Isopinocampheyl-9-borabicyclo[3.3.1]nonane
Molecular Weight (g/mol) 429.29 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.