Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich R-Alpine-Borane™ Solution

Catalog No. 232734100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-273-4100ML 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 23-273-4100ML Supplier Sigma Aldrich Supplier No. 232734100ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

R-Alpine-Borane™ is a chiral reducing agent, synthesized from (+)-+a-pinene via hydroboration.R-Alpine-Borane™ solution may be used to prepare:™-5-(Benzyloxy)pent-3-y-2-ol, an intermediate for the stereoselective total synthesis of (-)-stagonolide D.™-1-(p-Tolyl)-1-pentyn-3-ol, an intermediate for the enatioselective synthesis of ™-incrustoporin.N-(tert-Butyloxycarbonyl)-(4S)-[4-2H]-1-L-homoserine tert-butyl ester, an intermediate for synthesizing (4S)-[4-2H]-L-homoserine.Alpine-Borane is a registered trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 135540-11-3
Melting Point 153°C to 156°C
Molecular Formula C9H16ClF6N2P
Linear Formula C9H16N2Cl -+ PF6
MDL Number MFCD00191333
Quantity 5 g
Synonym N,N,N',N'-Bis(tetramethylene)chloroformamidinium hexafluorophosphate; Chloro-N,N,N',N'-bis(tetramethylene)formamidinium hexafluorophosphate; PyClU
Molecular Weight (g/mol) 332.65 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.