Learn More
Sigma Aldrich Fine Chemicals Biosciences (R)-(−)-α-Methoxy-α-(trifluoromethyl)phenylacetyl chloride, 39637-99-5, MFCD00044400, 500 mg
Catalog No. NC1203336
Encompass
Linear Formula: C6H5C(OCH3)(CF3)COCl, Molecular Weight: 252.62, for chiral derivatization, LiChropur, ≥99.0%, Synonym: (R)-(−)-MTPA-Cl, Mosher’s acid chloride. It was used for derivatizing D-arabinitol and L-arabinitol samples for high performance thin layer chromatography (HPTLC) analysis for studying resolution of the diastereoisomer. Ready-to-use reagent for the determination of the enantiomeric purity of alcohols and amines after derivatization.
Catalog No. NC1203336 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 65363500MG
Linear Formula: C6H5C(OCH3)(CF3)COCl, Molecular Weight: 252.62, for chiral derivatization, LiChropur, ≥99.0%, Synonym: (R)-(−)-MTPA-Cl, Mosher’s acid chloride. It was used for derivatizing D-arabinitol and L-arabinitol samples for high performance thin layer chromatography (HPTLC) analysis for studying resolution of the diastereoisomer. Ready-to-use reagent for the determination of the enantiomeric purity of alcohols and amines after derivatization.