Learn More
Medchemexpress LLC (R)-aminocarnitine (emeriamine) | 98063-21-9 | MFCD27977645 | 100.0% | 160.21 g/mol | C7H16N2O2 | 100MG
SDP
Catalog No. 5000239621
Encompass_Preferred
(R)-Aminocarnitine (emeriamine) is a research reagent that inhibits fatty acid oxidation and is used to study metabolic disease, lipid metabolism, and related biochemical pathways. It has formula C7H16N2O2, molecular weight 160.21 g/mol, and is supplied with high reported purity for laboratory research.
  • Inhibits fatty acid oxidation.
  • Reduces hyperglycemia and ketosis in reported studies.
  • Used for research into metabolic disease and energy metabolism.
  • Reported high purity suitable for biochemical assays.
  • Supplied in a range of small package sizes for laboratory use.
Catalog No. 50-002-39621 Supplier Medchemexpress LLC Supplier No. HY126221A100MG
(R)-Aminocarnitine (emeriamine) is a research reagent that inhibits fatty acid oxidation and is used to study metabolic disease, lipid metabolism, and related biochemical pathways. It has formula C7H16N2O2, molecular weight 160.21 g/mol, and is supplied with high reported purity for laboratory research.
  • Inhibits fatty acid oxidation.
  • Reduces hyperglycemia and ketosis in reported studies.
  • Used for research into metabolic disease and energy metabolism.
  • Reported high purity suitable for biochemical assays.
  • Supplied in a range of small package sizes for laboratory use.