Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-Amlodipine | 103129-81-3 | MFCD06809924 | 99.5% | 408.88 g/mol | C20H25ClN2O5 | 10 MG
SDP

Catalog No. 5000298891
Encompass_Preferred

(R)-Amlodipine is the R-enantiomer of amlodipine, a dihydropyridine L-type calcium channel inhibitor supplied as an off-white to light yellow solid for research use. It is commonly used in pharmacological and formulation studies that require the isolated enantiomer.

  • High purity (99.45%).
  • Molecular weight 408.88 g/mol.
  • Chemical formula C20H25ClN2O5.
  • Soluble in DMSO at 100 mg/mL; ultrasonic recommended.
  • In vivo formulation protocols yield ≥2.5 mg/mL solubility in listed vehicles.
  • Available in small research sizes (e.g., 10 mg).
  • Storage: powder -20°C for up to 3 years; in solvent -80°C for 6 months or -20°C for 1 month.

Catalog No. 50-002-98891 Supplier Medchemexpress LLC Supplier No. HYB0317D10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R)-Amlodipine is the R-enantiomer of amlodipine, a dihydropyridine L-type calcium channel inhibitor supplied as an off-white to light yellow solid for research use. It is commonly used in pharmacological and formulation studies that require the isolated enantiomer.

  • High purity (99.45%).
  • Molecular weight 408.88 g/mol.
  • Chemical formula C20H25ClN2O5.
  • Soluble in DMSO at 100 mg/mL; ultrasonic recommended.
  • In vivo formulation protocols yield ≥2.5 mg/mL solubility in listed vehicles.
  • Available in small research sizes (e.g., 10 mg).
  • Storage: powder -20°C for up to 3 years; in solvent -80°C for 6 months or -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.