Learn More
Medchemexpress LLC (R)-Amlodipine-d4 | 1346616-97-4 | 500 UG
SDP
Catalog No. 5000406735
Encompass_Preferred
(R)-Amlodipine-d4 is a deuterium-labeled form of (R)-Amlodipine. Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules for use as tracers in drug development for quantitation. Deuteration has gained attention due to its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium-labeled form of (R)-Amlodipine
  • Stable heavy isotopes for use as tracers
  • Potential to influence pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-06735 Supplier Medchemexpress LLC Supplier No. HYB0317DS500UG
(R)-Amlodipine-d4 is a deuterium-labeled form of (R)-Amlodipine. Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules for use as tracers in drug development for quantitation. Deuteration has gained attention due to its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium-labeled form of (R)-Amlodipine
  • Stable heavy isotopes for use as tracers
  • Potential to influence pharmacokinetic and metabolic profiles of drugs