Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-Apremilast | 608141-44-2 | 100.0% | 460.50 g/mol | C22H24N2O7S | 10 MG
SDP

Catalog No. 5000297775
Encompass_Preferred

(R)-Apremilast is the R-enantiomer of apremilast provided as a high-purity analytical standard for research use only. It is supplied as a white to yellow solid and can be provided as a solid or as a 10 mM solution in DMSO; safety and analytical documentation are available.

  • Purity 99.99% (analytical grade).
  • CAS number 608141-44-2.
  • Molecular formula C22H24N2O7S.
  • Molecular weight 460.50 g/mol.
  • Available as solids (5 mg-500 mg) and 10 mM DMSO solutions.
  • Intended for research use only, not for human or animal therapeutic use.
  • Datasheet, COA, and SDS available for handling and analysis.

Catalog No. 50-002-97775 Supplier Medchemexpress LLC Supplier No. HYI063610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R)-Apremilast is the R-enantiomer of apremilast provided as a high-purity analytical standard for research use only. It is supplied as a white to yellow solid and can be provided as a solid or as a 10 mM solution in DMSO; safety and analytical documentation are available.

  • Purity 99.99% (analytical grade).
  • CAS number 608141-44-2.
  • Molecular formula C22H24N2O7S.
  • Molecular weight 460.50 g/mol.
  • Available as solids (5 mg-500 mg) and 10 mM DMSO solutions.
  • Intended for research use only, not for human or animal therapeutic use.
  • Datasheet, COA, and SDS available for handling and analysis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.