Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-Azetidine-2-carboxylic acid | 7729-30-8 | MFCD00800731 | 99.6% | 101.10 g/mol | C4H7NO2 | 1 G
SDP

Catalog No. 5000203072
Encompass_Preferred

(R)-Azetidine-2-carboxylic acid is the R-enantiomer of azetidine-2-carboxylic acid, a nonproteinogenic amino acid used as an ADC linker and in biochemical research. The material is supplied as a solid for laboratory use.

  • R-enantiomer of azetidine-2-carboxylic acid.
  • Nonproteinogenic amino acid and ADC linker.
  • Molecular formula C4H7NO2; molecular weight 101.10 g/mol.
  • Purity reported as 99.6% (data sheet) and 99.9% (alternate listing).
  • Appearance solid; store protected from light at 4°C; in solution store at -80°C (6 months) or -20°C (1 month).
  • Available in small laboratory pack sizes for research use.

Catalog No. 50-002-03072 Supplier Medchemexpress LLC Supplier No. HYW0177551G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R)-Azetidine-2-carboxylic acid is the R-enantiomer of azetidine-2-carboxylic acid, a nonproteinogenic amino acid used as an ADC linker and in biochemical research. The material is supplied as a solid for laboratory use.

  • R-enantiomer of azetidine-2-carboxylic acid.
  • Nonproteinogenic amino acid and ADC linker.
  • Molecular formula C4H7NO2; molecular weight 101.10 g/mol.
  • Purity reported as 99.6% (data sheet) and 99.9% (alternate listing).
  • Appearance solid; store protected from light at 4°C; in solution store at -80°C (6 months) or -20°C (1 month).
  • Available in small laboratory pack sizes for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.