Learn More
Bellco Glass, Inc. Round Bottom Canister Only,38X430mm
SDP
Catalog No. 501944660
Encompass_Preferred
ROUND BOTTOM GLASS PIPETTE CANISTER. MANUFACTURED FROM TYPE 1, CLASS A BOROSILICATE GLASS COMPONENTS WILL WITHSTAND REPEATED DRY OR STEAM HEAT STERILIZATION MADE IN THE USA CASE Qty: 6
Catalog No. 50-194-4660 Supplier Bellco Glass, Inc. Supplier No. 128700430
ROUND BOTTOM GLASS PIPETTE CANISTER. MANUFACTURED FROM TYPE 1, CLASS A BOROSILICATE GLASS COMPONENTS WILL WITHSTAND REPEATED DRY OR STEAM HEAT STERILIZATION MADE IN THE USA CASE Qty: 6