Learn More
StatLab R.B.D. Decal Solution
SDP
Catalog No. 22000033
Change view
Click to view available options
Solution Type:
Decalcifier
Catalog No. Solution Type
22-000-033 Decalcifier
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-000-033 Supplier StatLab Supplier No. DCR0257
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
A fast and aggressive decal solution that will decalcify most bone marrow biopsies in 30 minutes
  • Large bone sections will decalcify in 2 to 6 hours
  • Readily biodegradable and non-corrosive
  • Will distinguish chromatin and show superior surface decalcification
  • The perfect choice for most routine work
Specifications
Product Type Decalcifying Solution
Solution Type Decalcifier
Form Solution