Learn More
Bellco Glass, Inc. Round Bottom Flange Vessel,10L
SDP
Catalog No. 501944894
Encompass_Preferred
10L bioreactor vessel 240 x 490mm, round bottom. Heavy walled construction for cell culture and fermentation applications. Heavy duty o-ring flange creates positive seal for stainless steel head-plate. For use with stainless steel head-plate, o-ring, and locking clamp. Use with CellTrol and speed control module. MADE IN THE USA.
Catalog No. 50-194-4894 Supplier Bellco Glass, Inc. Supplier No. 196488410
10L bioreactor vessel 240 x 490mm, round bottom. Heavy walled construction for cell culture and fermentation applications. Heavy duty o-ring flange creates positive seal for stainless steel head-plate. For use with stainless steel head-plate, o-ring, and locking clamp. Use with CellTrol and speed control module. MADE IN THE USA.