Learn More
Bellco Glass, Inc. Round Bottom Marine Impeller,15L
SDP
Catalog No. 501944885
Encompass_Preferred
MADE IN THE USA. Blade dimensions: 5 3/4" x 2"Shaft: 14 1/2"Paddles and impeller assemblies for cell cultureConstructed of 316l electropolished stainless steelReduces shear in delicate cell cultureMarine impeller includes 1/2" shaft and impeller bladeOmni and z-z paddles are one piece welded, 1/4" shaft
Catalog No. 50-194-4885 Supplier Bellco Glass, Inc. Supplier No. 196488152
MADE IN THE USA. Blade dimensions: 5 3/4" x 2"Shaft: 14 1/2"Paddles and impeller assemblies for cell cultureConstructed of 316l electropolished stainless steelReduces shear in delicate cell cultureMarine impeller includes 1/2" shaft and impeller bladeOmni and z-z paddles are one piece welded, 1/4" shaft