Learn More
Bellco Glass, Inc. Round Bottom Marine Impeller,15L
SDP
Supplier:  Bellco Glass, Inc. 196473015
Encompass_Preferred
MADE IN THE USA. Blade dimension: 4" od Marine prop – Right-HandedShaft oal:14 1/2"Paddles and impeller assemblies for cell cultureConstructed of 316l electropolished stainless steelReduces shear in delicate cell cultureMarine impeller includes 1/2" shaft and impeller bladeOmni and z-z paddles are one piece welded, 1/4" shaft
Catalog No. 50-194-4862