Learn More
Bellco Glass, Inc. Round Bottom Marine Impeller,36L
SDP
Catalog No. 501944863
Encompass_Preferred
MADE IN THE USA. Blade dimension: 4"OD Marine prop – Right-HandedShaft OAL: 16 1/4Paddles and impeller assemblies for cell cultureConstructed of 316l electropolished stainless steelReduces shear in delicate cell cultureMarine impeller includes 1/2" shaft and impeller bladeOmni and z-z paddles are one piece welded, 1/4" shaft
Catalog No. 50-194-4863 Supplier Bellco Glass, Inc. Supplier No. 196473036
MADE IN THE USA. Blade dimension: 4"OD Marine prop – Right-HandedShaft OAL: 16 1/4Paddles and impeller assemblies for cell cultureConstructed of 316l electropolished stainless steelReduces shear in delicate cell cultureMarine impeller includes 1/2" shaft and impeller bladeOmni and z-z paddles are one piece welded, 1/4" shaft