Learn More
Bellco Glass, Inc. Round Bottom Marine Impeller,5L Complete
SDP
Supplier:  Bellco Glass, Inc. 196473005
Encompass_Preferred
Dimensions: 1/2" od x 7 1/2" longStainless steel 4" od right hand pitch marine prop & e-clipMaterial: 316l stainless steel with electropolish finishFor round bottom omni vessel (1964-88405)Need l/f head plate, over head drive motor Size: 5l MADE IN THE USA.
Catalog No. 50-194-4860