Learn More
Bellco Glass, Inc. Round Bottom Marine Impeller,8L
SDP
Catalog No. 501944861
Encompass_Preferred
MADE IN THE USA. Dimensions: 1/2" od x 12 1/2" longStainless steel 4" od right hand pitch marine prop & e-clipMaterial: 316l stainless steel with electropolish finishFor round bottom omni vessel (1964-88405)Need l/f head plate, over head drive motorSize: 8l
Catalog No. 50-194-4861 Supplier Bellco Glass, Inc. Supplier No. 196473008
MADE IN THE USA. Dimensions: 1/2" od x 12 1/2" longStainless steel 4" od right hand pitch marine prop & e-clipMaterial: 316l stainless steel with electropolish finishFor round bottom omni vessel (1964-88405)Need l/f head plate, over head drive motorSize: 8l