Learn More
Medchemexpress LLC (R)-Baclofen (Arbaclofen) | 69308-37-8 | 99.50% | 213.66 | 200 MG
SDP
Catalog No. 5000385677
Encompass_Preferred
R-Baclofen (Arbaclofen) is a selective GABAB receptor agonist commonly used in research. It exhibits agonist activity at human GABA-B B1/B2 receptors expressed in HEK293 cells, as assessed by the inhibition of forskolin-stimulated cAMP accumulation. In vivo studies demonstrate its ability to suppress audiogenic seizures in Fmr1-knockout mice and significantly reduce seizure incidence with acute administration.
  • Selective GABAB receptor agonist.
  • Exhibits agonist activity at human GABA-B B1/B2 receptors.
  • Suppresses audiogenic seizures in Fmr1-knockout mice.
  • Reduces seizure incidence in mice.
Catalog No. 50-003-85677 Supplier Medchemexpress LLC Supplier No. HY17354200MG
R-Baclofen (Arbaclofen) is a selective GABAB receptor agonist commonly used in research. It exhibits agonist activity at human GABA-B B1/B2 receptors expressed in HEK293 cells, as assessed by the inhibition of forskolin-stimulated cAMP accumulation. In vivo studies demonstrate its ability to suppress audiogenic seizures in Fmr1-knockout mice and significantly reduce seizure incidence with acute administration.
  • Selective GABAB receptor agonist.
  • Exhibits agonist activity at human GABA-B B1/B2 receptors.
  • Suppresses audiogenic seizures in Fmr1-knockout mice.
  • Reduces seizure incidence in mice.