Learn More
Enzo Life Sciences R-(+)-Baclofen. hydrochloride (50mg). CAS: 63701-55-3
SDP
Supplier:  Enzo Life Sciences ALX550074M050
Encompass_Preferred
Selective GABAB receptor agonist. Skeletal muscle relaxant. The more active enantiomer of (±)-Baclofen (BML-EA128). Alternative name: R-(+)-beta-(Aminomethyl)-4-chlorobenzenepropanoic acid . HCl, R-(+)-4-Amino-3-(4-chlorophenyl)butanoic acid . HCl. Purity: ≥98% (HPLC). Solubility: Soluble in water, dilute aqueous base or DMSO. Long Term Storage: +4°C.
Catalog No. 50-200-9474