Learn More
Abcam (R)-Baclofen (mM/ml), GABAB agonist, 25MM_ML
Catalog No. 502704511
Encompass_Preferred
MW 213.66 Da, Purity >98%. More active enantiomer of selective GABAB agonist (R,S)-Baclofen (ab120149). Soluble in 1 ml water to give specified mM/ml concentration.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4511 Supplier Abcam Supplier No. AB14671425MM_ML
MW 213.66 Da, Purity >98%. More active enantiomer of selective GABAB agonist (R,S)-Baclofen (ab120149). Soluble in 1 ml water to give specified mM/ml concentration.
The product is subject to the following: Abcam Restricted Use Statement