Learn More
Medchemexpress LLC (R)-BAY-85-8501 | 2446175-39-7 | 99.1% | 474.46 | 1 ML
SDP
Catalog No. 5000401505
Encompass_Preferred
(R)-BAY-85-8501 is the less active enantiomer of BAY-85-8501. BAY-85-8501 is a selective and potent inhibitor of Human Neutrophil Elastase (HNE), with an IC50 of 65 pM. It is intended for research use only.
  • Less active enantiomer of BAY-85-8501
  • Selective and potent inhibitor of human neutrophil elastase (HNE)
  • For research use only
Catalog No. 50-004-01505 Supplier Medchemexpress LLC Supplier No. HY19908B10MM/1ML
(R)-BAY-85-8501 is the less active enantiomer of BAY-85-8501. BAY-85-8501 is a selective and potent inhibitor of Human Neutrophil Elastase (HNE), with an IC50 of 65 pM. It is intended for research use only.
  • Less active enantiomer of BAY-85-8501
  • Selective and potent inhibitor of human neutrophil elastase (HNE)
  • For research use only