Learn More
Medchemexpress LLC HY-19908B 5mg Medchemexpress, (R)-BAY-85-8501 CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY19908B5MG
Encompass_Preferred
Medchemexpress, HY-19908B 5mg (R)-BAY-85-8501 CAS: (R)-BAY-85-8501 is the less active Enantiomer of BAY-85-8501. BAY-85-8501 is a selective and potent inhibitor of Human Neutrophil Elastase (HNE) , with an IC 50 of 65 pM [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2844