Learn More
Medchemexpress LLC (R)-Bay-899 | 2988191-40-6 | 99.9% | 459.45 | C25H19F2N5O2 | 5 MG
SDP
Catalog No. 5000262759
Encompass_Preferred
(R)-BAY-899 is the R-enantiomer of BAY-899, an orally active, selective luteinizing hormone receptor (LH-R) antagonist used as a research tool for in vitro and in vivo studies. Supplied as an off-white to light yellow solid, the compound is characterized by high purity and includes documented solubility and storage conditions for laboratory applications.
  • Selective LH-R antagonist for receptor signaling studies.
  • High purity suitable for analytical and biological assays.
  • Documented solubility in DMSO and multiple in vivo formulations.
  • Stable powder storage at -20°C or 4°C with defined solvent stability.
  • Available in small solid and solution pack sizes for research use.
Catalog No. 50-002-62759 Supplier Medchemexpress LLC Supplier No. HY130248B5MG
(R)-BAY-899 is the R-enantiomer of BAY-899, an orally active, selective luteinizing hormone receptor (LH-R) antagonist used as a research tool for in vitro and in vivo studies. Supplied as an off-white to light yellow solid, the compound is characterized by high purity and includes documented solubility and storage conditions for laboratory applications.
  • Selective LH-R antagonist for receptor signaling studies.
  • High purity suitable for analytical and biological assays.
  • Documented solubility in DMSO and multiple in vivo formulations.
  • Stable powder storage at -20°C or 4°C with defined solvent stability.
  • Available in small solid and solution pack sizes for research use.